Gallium Standard: Ga @ 1000 µg/mL in 5% HNO3, 0.5% HCl, VHG, LGC Standards
Supplier: LGC Standards
VHG-PGANH-100
77697-759EA 56.2 USD
77697-759
Gallium Standard: Ga @ 1000 µg/mL in 5% HNO3, 0.5% HCl, VHG, LGC Standards
Standards Cationic Standards Gallium Standards