Nickel (II) nitrate 1% Ni(NO3)2 (w/v) in nitric acid 2% matrix modifier, for graphite furnace AAS (CRM), VHG

Supplier: LGC Standards
Warning

Synonyms: Nickel dinitrate

VHG-MNIN1P-100
77697-695EA 82.85 USD
77697-695
Nickel (II) nitrate 1% Ni(NO3)2 (w/v) in nitric acid 2% matrix modifier, for graphite furnace AAS (CRM), VHG
Nickel (II) nitrate

This solution is intended for use as a matrix modifier CRM for graphite furnace atomic absorption spectroscopy (GFAA).


Caution: VHG ensures the accuracy of this solution for 12 Months from the Certification date.

Formula: N₂NiO₆
MW: 182.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011139
CAS Number: 13138-45-9

Order Now

Specification Test Results

Nickel Nitrate Ni(NO3)2 10,000 ± 50 µg/m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR