Magnesium nitrate 1% Mg(NO₃)₂ (w/v) in nitric acid 2% matrix modifier, for graphite furnace AAS (CRM), VHG

Supplier: LGC Standards
Warning

VHG-MMGN1P-100
77697-694EA 82.85 USD
77697-694
Magnesium nitrate 1% Mg(NO₃)₂ (w/v) in nitric acid 2% matrix modifier, for graphite furnace AAS (CRM), VHG
Magnesium nitrate solutions

This solution is intended for use as a matrix modifier CRM for graphite furnace atomic absorption spectroscopy (GFAA).


Caution: VHG ensures the accuracy of this solution for 12 Months from the Certification date.

Formula: Mg(NO₃)₂
CAS Number: 10377-60-3

Order Now

Specification Test Results

Magnesium Nitrate (Mg(NO3)2) 10,000 ± 50 µg/mL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR