Scandium Standard: Sc @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Supplier: LGC Standards
VHG-LSCN-100
77697-668EA 139.05 USD
77697-668
Scandium Standard: Sc @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Standards Cationic Standards Scandium Standards