Gold Au @ 100 µg/mL in hydrochloric acid 5% stabilizer for mercury, reference material (CRM), VHG

Supplier: LGC Standards
Warning

VHG-LSAU-100
77697-667EA 141.85 USD
77697-667
Gold Au @ 100 µg/mL in hydrochloric acid 5% stabilizer for mercury, reference material (CRM), VHG
Gold

This solution is intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).


Caution: VHG ensures the accuracy of this solution for 12 Months from the Certification date.

Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Au 100.0 ± 0.5 µg/mL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR