Lubricating Oil Standard 6: Ca @ 0.400; P @ 0.200; S @ 0.175; Zn @ 0.050 wt% in Lubricating Oil, VHG, LGC Standards
Supplier: LGC Standards
VHG-LOIL6-100
77697-659EA 386.75 USD
77697-659
Lubricating Oil Standard 6: Ca @ 0.400; P @ 0.200; S @ 0.175; Zn @ 0.050 wt% in Lubricating Oil, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards