Major Elements Standard (Primary): Ca, Fe, K, Mg, Na @ 2000 µg/mL in 5% HNO3, VHG, LGC Standards
Supplier: LGC Standards
VHG-LMES-100
77697-639EA 211.45 USD
77697-639
Major Elements Standard (Primary): Ca, Fe, K, Mg, Na @ 2000 µg/mL in 5% HNO3, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards