Internal Standard Multi-Element Mix 4: In, Tb, Bi @ 10; Ge, Te @ 25; 6Li, Sc @ 50 µg/mL in 5% HNO3, tr. F-, VHG, LGC Standards
Supplier: LGC Standards
VHG-LIS4-100
77697-623EA 185.65 USD
77697-623
Internal Standard Multi-Element Mix 4: In, Tb, Bi @ 10; Ge, Te @ 25; 6Li, Sc @ 50 µg/mL in 5% HNO3, tr. F-, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards