Internal Standard Multi-Element Mix 3: 6Li, Sc, Ge, In, Tb, Lu, Bi @ 100 µg/mL in 5% HNO3, tr. F-, VHG, LGC Standards
Supplier: LGC Standards
VHG-LIS3-100
77697-622EA 191.45 USD
77697-622
Internal Standard Multi-Element Mix 3: 6Li, Sc, Ge, In, Tb, Lu, Bi @ 100 µg/mL in 5% HNO3, tr. F-, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards