Multi-Anion Standard 4: F- @ 100; Cl- @ 200; Br-, NO3-, SO4(-2) @ 400; PO4(-3) @ 600 µg/mL in H2O, VHG, LGC Standards
Supplier: LGC Standards
VHG-ICM4-100
77697-546EA 171.35 USD
77697-546
Multi-Anion Standard 4: F- @ 100; Cl- @ 200; Br-, NO3-, SO4(-2) @ 400; PO4(-3) @ 600 µg/mL in H2O, VHG, LGC Standards
Standards Anionic Standards Multi-Element Anioinic Standards