Pure Water Density (Nominal: 0.99909 g/mL @ 15°C), Values at 15, 20 & 25°C, Paragon Scientific, LGC Standards
Supplier: LGC Standards
ALK-DEN-WAT
77696-029EA
146.65
USD
77696-029
Pure Water Density (Nominal: 0.99909 g/mL @ 15°C), Values at 15, 20 & 25°C, Paragon Scientific, LGC Standards
Standards
Density Standards