Multi-Element Four Component Standard: 4.0 wt% S, 350 µg/g Fe, 30 µg/g Ni, 200 µg/g V in 20 cSt Mineral Oil, VHG, LGC Standards
Supplier: LGC Standards
VHG-SMOIL6-100
77695-592EA 384.6 USD
77695-592
Multi-Element Four Component Standard: 4.0 wt% S, 350 µg/g Fe, 30 µg/g Ni, 200 µg/g V in 20 cSt Mineral Oil, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards