Sulfur Standard: S @ 7500 µg/g (0.750 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Supplier: LGC Standards
VHG-SMIN-7500-100
77695-583EA 102.85 USD
77695-583
Sulfur Standard: S @ 7500 µg/g (0.750 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Standards Cationic Standards Sulfur standards