Sulfur Standard: S @ 1000 ug/g (0.1 wt%) in 80% Isooctane/20% Toluene, VHG, LGC Standards
Supplier: LGC Standards
VHG-SISO80TOL20-1000-100
77695-536EA 161.95 USD
77695-536
Sulfur Standard: S @ 1000 ug/g (0.1 wt%) in 80% Isooctane/20% Toluene, VHG, LGC Standards
Standards Cationic Standards Sulfur standards