Sulfur Standard: S @ 10000 µg/g (1.00 wt%) in #2 Diesel Fuel, VHG, LGC Standards
Supplier: LGC Standards
VHG-SDSL-1P-100
77695-484EA 108.55 USD
77695-484
Sulfur Standard: S @ 10000 µg/g (1.00 wt%) in #2 Diesel Fuel, VHG, LGC Standards
Standards Cationic Standards Sulfur standards