Titanium Standard: Ti @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Supplier: LGC Standards
VHG-OTI-5000-50G
77695-373EA 77.2 USD
77695-373
Titanium Standard: Ti @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Standards Cationic Standards Titanium Standards