Ammonium phosphate 10% (w/v) in nitric acid 2% matrix modifier, for graphite furnace AAS (CRM), VHG

Supplier: LGC Standards

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

VHG-MAP10P-100
77693-168EA 116.15 USD
77693-168
Ammonium phosphate 10% (w/v) in nitric acid 2% matrix modifier, for graphite furnace AAS (CRM), VHG
Ammonium dihydrogen phosphate

This solution is intended for use as a matrix modifier CRM for graphite furnace atomic absorption spectroscopy (GFAA).


Caution: VHG ensures the accuracy of this solution for 12 Months from the Certification date.

Formula: NH₄H₂PO₄
MW: 115.03 g/mol
MDL Number: MFCD00003396
CAS Number: 7722-76-1

Order Now

Specification Test Results

Ammonium Phosphate (NH4H2PO4) 100,000 ± 500 µg/mL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR