Bismuth Internal Standard: Bi @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Supplier: LGC Standards
VHG-LISABI-100
77693-161EA 60 USD
77693-161
Bismuth Internal Standard: Bi @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Standards Cationic Standards Bismuth Standards