Multi-Element 18 Components 200.8 Calibration Standard A2: 10 µg/mL Al, As, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Th, Tl, U, V, Zn; 50 µg/mL Se in 5% HNO3, tr. Tartaric Acid, VHG, LGC Standards
Supplier: LGC Standards
VHG-L54SSA2-100
77693-148EA 259.9 USD
77693-148
Multi-Element 18 Components 200.8 Calibration Standard A2: 10 µg/mL Al, As, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Th, Tl, U, V, Zn; 50 µg/mL Se in 5% HNO3, tr. Tartaric Acid, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards