Multi-Element 18 Components 200.8 Calibration Standard A1: 10 µg/mL Al, As, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn in 5% HNO3, tr. Tartaric Acid, VHG, LGC Standards
Supplier: LGC Standards
VHG-L53SSA1-100
77693-147EA 254.2 USD
77693-147
Multi-Element 18 Components 200.8 Calibration Standard A1: 10 µg/mL Al, As, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn in 5% HNO3, tr. Tartaric Acid, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards