CLP Instrument Calibration Standard 1: Ag @ 200; Be @ 400; Cd @ 500; Ba, Co, Cu, Fe, Mn, Ni, Pb, Tl, Zn @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Supplier: LGC Standards
VHG-ICL1-100
77693-119EA 240.85 USD
77693-119
CLP Instrument Calibration Standard 1: Ag @ 200; Be @ 400; Cd @ 500; Ba, Co, Cu, Fe, Mn, Ni, Pb, Tl, Zn @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards