200.7 Rev. 4.4 - Calibration Standard 2: Sn @ 200; Li, Sb @ 500; Al, Fe, Mo, Na, SiO2, Ti @ 1000 µg/mL in 20% HCl, tr. HF, VHG, LGC Standards
Supplier: LGC Standards
VHG-44CS2Z-100
77692-993EA 214.2 USD
77692-993
200.7 Rev. 4.4 - Calibration Standard 2: Sn @ 200; Li, Sb @ 500; Al, Fe, Mo, Na, SiO2, Ti @ 1000 µg/mL in 20% HCl, tr. HF, VHG, LGC Standards
Standards Cationic Standards Multi-Element Standards