Density, Diesel (Nominal: 0.83418 g/mL @ 15°C), Paragon Scientific, LGC Standards
Supplier: LGC Standards
ALK-CRMU-DEGO
77691-403EA
241.9
USD
77691-403
Density, Diesel (Nominal: 0.83418 g/mL @ 15°C), Paragon Scientific, LGC Standards
Standards
Density Standards