HardyDisks™ Hippurate Disks
Supplier: HARDY DIAGNOSTICS
Z18
77691-386PK 43.36 USD
77691-386
HardyDisks™ Hippurate Disks
Microbiology Test Systems Antibiotic Tests
Hippurate disks are used for the sodium hippurate hydrolysis test. Glycine, an end product of hippurate hydrolysis, reacts with ninhydrin to produce a purple color. Hippurate is commonly used to presumptively identify Gardnerella vaginalis, Listeria monocytogenes, Campylobacter jejuni, and group B streptococci.
Product is light, temperature, and moisture sensitive.