Anti-ASF1A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A72069-100UL
77691-146EA 334.95 USD
77691-146
Anti-ASF1A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:200-1:1000, IHC:1:20-1:200

Type:
Antigen: ASF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR