Anti-INO80 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A71845-100UG
77691-122EA 334.95 USD
77691-122
Anti-INO80 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IF:1:50-1:200

Type:
Antigen: INO80
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR