Anti-CLEC4M Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A66729-100UG
77690-597EA 334.95 USD
77690-597
Anti-CLEC4M Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IF:1:50-1:200

Type:
Antigen: CLEC4M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR