Anti-C4orf3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62243-100UG A62243-50UG
77690-190EA 334.95 USD
77690-190 77719-917
Anti-C4orf3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000

Type:
Antigen: C4orf3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR