Anti-SULF1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A61100-100UG A61100-50UG
77690-072EA 334.95 USD
77690-072 77717-768
Anti-SULF1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IHC:1:100-1:500, IF:1:50-1:500

Type:
Antigen: SULF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR