Anti-SLC22A9 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60934-100UG A60934-50UG
77690-056EA 334.95 USD
77690-056 77717-452
Anti-SLC22A9 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: SLC22A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR