Anti-AMIGO2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A59431-100UG
77689-957EA 334.95 USD
77689-957
Anti-AMIGO2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IHC:1:500-1:1000, IF:1:200-1:500

Type:
Antigen: AMIGO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR