Anti-NOTUM Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A58456-100UG
77689-853EA 334.95 USD
77689-853
Anti-NOTUM Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: NOTUM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR