Anti-N Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A50315-100UG
77689-330EA 334.95 USD
77689-330
Anti-N Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: N
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Junin mammarenavirus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR