Anti-TRAF6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A37397-100UG
77688-828EA 334.95 USD
77688-828
Anti-TRAF6 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TRAF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR