Anti-TNNI2 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A37177-100UL
77688-810EA 334.95 USD
77688-810
Anti-TNNI2 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TNNI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR