Anti-RAF1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A32764-100UG
77688-551EA 334.95 USD
77688-551
Anti-RAF1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RAF1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR