Anti-RAD51 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A32756-100UG
77688-550EA 334.95 USD
77688-550
Anti-RAD51 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RAD51
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR