Anti-PNLIPRP3 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A31514-100UG
77688-455EA 334.95 USD
77688-455
Anti-PNLIPRP3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200

Type:
Antigen: PNLIPRP3
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR