Anti-PECR Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A30938-100UL
77688-411EA 334.95 USD
77688-411
Anti-PECR Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: PECR
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR