Anti-NMBR Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A29768-100UG
77688-336EA 334.95 USD
77688-336
Anti-NMBR Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: NMBR
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR