Anti-MYF6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A29202-100UG
77688-301EA 334.95 USD
77688-301
Anti-MYF6 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: MYF6
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR