4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl 98%

Supplier: Ambeed
Warning

A830397-1G A830397-5G
77159-394EA 50 USD
77159-394 77686-855
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl 98%
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl

Caution: for Research Use Only

Formula: C₁₃H₂₂NO₃
MDL Number: MFCD17677357
CAS Number: 15051-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR