3,5-Dichlorophenol 98%

Supplier: Ambeed
Danger

A676491-25G
103840-524EA 10 USD
103840-524 77686-644
3,5-Dichlorophenol 98%
3,5-Dichlorophenol

Caution: Inert atmosphere, room temperature

Formula: C₆H₄Cl₂O
Boiling Pt: 233 °C (760 mm Hg)
Melting Pt: 65…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002259
CAS Number: 591-35-5
UN: 2020
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR