Boc-D-Phe-OSu ≥98%

Supplier: Ambeed
Warning

Synonyms: Boc-D-phenylalanine N-hydroxysuccinimide ester, (R)-2,5-Ddioxopyrrolidin-1-yl 2-((tert-butoxycarbonyl)amino)-3-phenylpropanoate

A604638-5G
77686-529EA 41 USD
77686-529
Boc-D-Phe-OSu ≥98%
Boc-D-Phe-OSu

Caution: for Research Use Only

Formula: C₁₈H₂₂N₂O₆
MW: 362.38 g/mol
MDL Number: MFCD00270515
CAS Number: 3674-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR