[RuCl(p-cymene)((R)-binap)]Cl ≥98%

Supplier: Ambeed
Danger

Synonyms: Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-binap)]Cl, (R)-RuCl[(p-cymene)(BINAP)]Cl, Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, [(R)-Binap RuCl p-cymene]Cl

A427739-5G A427739-1G
77686-172EA 174 USD
77686-172 77686-171
[RuCl(p-cymene)((R)-binap)]Cl ≥98%
[RuCl(p-cymene)((R)-binap)]Cl

Caution: for Research Use Only

Formula: C₅₄H₄₆Cl₂P₂Ru
MDL Number: MFCD00134456
CAS Number: 145926-28-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR