2,5-Dimercapto-1,3,4-thiadiazole ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,3,4-Thiadiazole-2,5-dithiol
A305906-100G
A305906-500G
A305906-25G
77616-318EA
43.26
USD
77616-318
77616-320
77685-872
2,5-Dimercapto-1,3,4-thiadiazole ≥98%
2,5-Dimercapto-1,3,4-thiadiazole
Caution: for Research Use Only
Formula:
C₂H₂N₂S₃ MW: 150.25 g/mol |
MDL Number:
MFCD00003103 CAS Number: 1072-71-5 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...