Isoliquiritigenin 98+%

Supplier: Ambeed
Warning

A289179-100MG A289179-10MG A289179-1G A289179-250MG A289179-50MG A289179-1MG A289179-5G
77114-224EA 19.57 USD
77114-224 77114-226 77114-228 77114-230 77114-232 77685-826 77685-827
Isoliquiritigenin 98+%
Isoliquiritigenin

Caution: for Research Use Only

Formula: C₁₅H₁₂O₄
MDL Number: MFCD00075907
CAS Number: 961-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR