Fmoc-Lys(palmitoyl)-OH 98%

Supplier: Ambeed
Warning

A268188-5G A268188-1G A268188-25G
77153-578EA 89 USD
77153-578 77685-765 77892-514
Fmoc-Lys(palmitoyl)-OH 98%
Fmoc-Lys(palmitoyl)-OH

Caution: for Research Use Only

Formula: C₃₇H₅₄N₂O₅
MDL Number: MFCD00237026
CAS Number: 201004-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR