EMD-1214063 ≥98%

Supplier: Ambeed
Warning

A242274-10MG A242274-1G A242274-5MG A242274-100MG A242274-250MG
77685-678EA 45 USD
77685-678 77685-679 77685-680 77901-920 77901-750
EMD-1214063 ≥98%
EMD-1214063

Caution: for Research Use Only

Formula: C₂₉H₂₈N₆O₂
MDL Number: MFCD18452823
CAS Number: 1100598-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR