Cabozantinib 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide, XL184

A200488-100MG A200488-10G A200488-5MG A200488-10MG A200488-1G A200488-1MG A200488-5G A200488-250MG
77152-132EA 17 USD
77152-132 77152-134 77152-136 77550-280 77685-499 77685-500 77685-501 77892-272
Cabozantinib 98%
Cabozantinib

Caution: for Research Use Only

Formula: C₂₈H₂₄FN₃O₅
MDL Number: MFCD20926324
CAS Number: 849217-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR